missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Immersion Filter

Immersion Filter for reverse filtration.

Brand:  ROBU VitraPOR™ 24202

Additional Details : Weight : 0.11970kg

 View more versions of this product

Product Code. 16843056

  • 1410.00 SEK / Pack of 5

Estimated Shipment: 31-05-2022
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass 3.3
20 mm
2
Non-sterile
150 mm
Videos
SDS
Documents
Certificates
Special Offers

Special Offers