missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Immersion Filter

Immersion Filter for reverse filtration.

Brand:  ROBU VitraPOR™ 24203

Additional Details : Weight : 0.11970kg

 View more versions of this product

Product Code. 16853056

  • 1830.00 SEK / Pack of 5
Estimated Shipment: 28-06-2024
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass (3.3)
3
20 mm
Non-sterile
150 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers