Learn More
ROBU VitraPOR™ Immersion Filter
Immersion Filter for reverse filtration.
Brand: ROBU VitraPOR™ 24204
Additional Details : Weight : 0.11970kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass 3.3 | |
20 mm | |
4 |
Non-sterile | |
150 mm |