Learn More
ROBU VitraPOR™ Immersion Filter
Immersion Filter for reverse filtration.
Brand:  ROBU VitraPOR™ 24351
Additional Details : Weight : 0.13200kg
 View more versions of this product
Product Code. 16883056
  • 945.00 SEK / Pack of 2
Estimated Shipment: 28-06-2024
to see stock.
Add to basket
Description
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass (3.3)
Non-sterile
210 mm
1 μm
1
35 mm