missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12750-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes the human homolog of the yeast Imp3 protein. The protein localizes to the nucleoli and interacts with the U3 snoRNP complex. The protein contains an S4 domain.Specifications
IMP3 | |
Polyclonal | |
Unconjugated | |
IMP3 | |
BRMS2, C15orf12, IMP3, MRPS4, U3 snoRNP protein IMP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
102462, 315697, 55272 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q921Y2, Q9NV31 | |
Imp3 | |
IMP3 Fusion Protein Ag3477 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |