Learn More
IMP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16205-1-AP-20UL
 View more versions of this product
Product Code. 16895744
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
IMP4 forms a ternary complex with IMP3.
Specifications
Specifications
IMP4
Polyclonal
Unconjugated
IMP4
BXDC4, IMP4, U3 snoRNP protein IMP4
Rabbit
Antigen Affinity Chromatography
RUO
27993, 316317, 92856
-20°C
Liquid
Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5PQR5, Q8VHZ7, Q96G21
Imp4
IMP4 Fusion Protein Ag9175
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IMP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated