Learn More
INA Mouse anti-Human, Unconjugated, Clone: 3C12A8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67120-1-IG-150UL
 View more versions of this product
Product Code. 16879565
  • 4652.90 SEK / 150µL
Description
The INA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human INA. This antibody recognizes human, mouse, rat, pig antigen. The INA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
INA
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, ELISA
3C12A8
Unconjugated
150 μL
9118
IgG2a
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Product Title
INA Mouse anti-Human, Unconjugated, Clone: 3C12A8, Proteintech > 150 μL