Learn More
INA Mouse anti-Human, Unconjugated, Clone: 3C12A8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67120-1-IG-150UL
Description
The INA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human INA. This antibody recognizes human, mouse, rat, pig antigen. The INA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
INA | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, Immunohistochemistry, ELISA | |
3C12A8 | |
Unconjugated | |
150 μL | |
9118 | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.