missing translation for 'onlineSavingsMsg'
Learn More

INE1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 21340-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16880945

  • 3665.00 SEK / 150µL
Estimated Shipment: 24-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

INE1
Polyclonal
Unconjugated
INE1
INE1, NCRNA00010
Rabbit
Antigen Affinity Chromatography
RUO
8552
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O15225
INE1
INE1 Fusion Protein Ag15601
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers