Learn More
INF2 Mouse anti-Human, Unconjugated, Clone: 4A11A7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66910-1-IG-150UL
 View more versions of this product
Product Code. 16879195
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The INF2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human INF2. This antibody recognizes human antigen. The INF2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
INF2
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
4A11A7
Unconjugated
150 μL
64423
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
INF2 Mouse anti-Human, Unconjugated, Clone: 4A11A7, Proteintech > 150 μL