Learn More
Lauda Coupler plug, 3 pins for contact input ''fault'' o
Brand:  Lauda EQS 048
326.84 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Coupler plug, 3 pins for contact input ''fault'' o
Product Code. 10043643
  • 360.00 SEK / Each
Product Title
Lauda Coupler plug, 3 pins for contact input ''fault'' o >