Learn More
INSR Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20433-1-AP-150UL
 View more versions of this product
Product Code. 16889964
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The insulin receptor (IR) is a heterodimeric protein complex that has an intracellular beta subunit and an extracellular alpha subunit, which is disulfide- linked to a transmembrane segment. The insulin ligand binds to the IR and initiates molecular signaling pathways that promote glucose uptake in cells and glycogen synthesis. Insulin binding to IR induces phosphorylation of intracellular tyrosine kinase domains and recruitment of multiple SH2 and SH3 domain-containing intracellular proteins that serve as signaling intermediates for pleiotropic effects of insulin.
Specifications
Specifications
INSR
Polyclonal
Unconjugated
INSR
CD220, HHF5, INSR, insulin receptor, IR
Rabbit
Antigen Affinity Chromatography
RUO
3643
-20°C
Liquid
Immunoprecipitation, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P06213
INSR
INSR Fusion Protein Ag13922
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
INSR Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated