Learn More
Internal Standard Mix for ICP-MS Systems, 8 Components, Honeywell Fluka™
Shop All Honeywell Fluka ProductsDescription
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Specifications
Chemical Name or Material | Internal Standard Mix for ICP-MS Systems |
Density | 1.06 g/cm3 |
pH | <2 |
Quantity | 100 mL |
Grade | Standards for ICP and ICP-MS |