missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Intersectin 1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21862-1-AP-150UL
This item is not returnable.
View return policy
Descripción
The protein encoded by this gene is one of the three catalytic subunits of protein phosphatase 1. PP1 is a serine/threonine specific protein phosphatase known to be involved in the regulation of a variety of cellular processes, such as cell division, glycogen metabolism, muscle contractility, protein synthesis, and HIV-1 viral transcription. Mouse studies suggest that PP1 functions as a suppressor of learning and memory. Two alternatively spliced transcript variants encoding distinct isoforms have been observed.Especificaciones
Intersectin 1 | |
Polyclonal | |
Unconjugated | |
ITSN1 | |
Intersectin 1, ITSN, ITSN1, SH3D1A, SH3P17 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6453 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15811 | |
ITSN1 | |
Intersectin 1 Fusion Protein Ag16517 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Intersectin 1 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido