missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INTU Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24914-1-AP-150UL
This item is not returnable.
View return policy
Description
Plays a key role in ciliogenesis and embryonic development. Regulator of cilia formation by controlling the organization of the apical actin cytoskeleton and the positioning of the basal bodies at the apical cell surface, which in turn is essential for the normal orientation of elongating ciliary microtubules. Plays a key role in definition of cell polarity via its role in ciliogenesis but not via conversion extension. Has an indirect effect on hedgehog signalingSpecifications
INTU | |
Polyclonal | |
Unconjugated | |
INTU | |
INT, INTU, KIAA1284, PDZD6, PDZK6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27152, 380614 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q059U7, Q9ULD6 | |
Intu | |
INTU Fusion Protein Ag19313 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
INTU Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction