missing translation for 'onlineSavingsMsg'
Learn More

Iodine Solution 1% (w/v) In Potassium Iodide, Pure, Fisher Chemical™

433.00 SEK - 940.00 SEK

Chemical Identifiers

MDL Number 11355
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10244282
SDS View Documents
Fisher Chemical
J/4450/15
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10122100
SDS View Documents
Fisher Chemical
J/4450/17
2.5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Iodine Solution 1% (w/v) In Potassium Iodide, Pure

Specifications

Chemical Identifiers

11355

Specifications

Amber
11355
Amber glass bottle
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Iodine solution 1% w/v in potassium iodide

Signal Word
  • Warning

Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodine Solution 1% (w/v) In Potassium Iodide, Pure, Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.