Learn More
IPO4 Mouse anti-Human, Unconjugated, Clone: 2F3H2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67549-1-IG-150UL
 View more versions of this product
Product Code. 16890236
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The IPO4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IPO4. This antibody recognizes human antigen. The IPO4 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
IPO4
Monoclonal
933 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
2F3H2
Unconjugated
150 μL
79711
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IPO4 Mouse anti-Human, Unconjugated, Clone: 2F3H2, Proteintech > 150 μL