Learn More
IPO4 Mouse anti-Human, Unconjugated, Clone: 2F3H2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67549-1-IG-150UL
 View more versions of this product
Product Code. 16890236
  • 4652.90 SEK / 150µL
Description
The IPO4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IPO4. This antibody recognizes human antigen. The IPO4 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
IPO4
Monoclonal
933 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
2F3H2
Unconjugated
150 μL
79711
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
IPO4 Mouse anti-Human, Unconjugated, Clone: 2F3H2, Proteintech > 150 μL