Learn More
Memmert™ Iq/Oq Document W/Device-Specific Works Test Data (Temperature And Humidity Value), For Ich110Eco
Includes temperature distribution survey for one free-selectable temperature and humidity value.
Brand: Memmert™ ICH110ecoD00136
Includes:
Works test data, temperature distribution survey
Description
- IQ/OQ document with works test data for chamber
- 27 measuring points to DIN 12880:2007-05, PQ check list as support for validation by customer (models HPP and ICH)
Specifications
110, 260, 750 Models Constant Climate Chamber | |
IQ/OQ Documentation | |
One Free-Selectable Temperature and Humidity Value for 27 Measuring Points to DIN 12880:2007-05 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Iq/Oq Document W/Device-Specific Works Test Data (Temperature And Humidity Value), For Ich110Eco >
Spot an opportunity for improvement?Share a Content Correction