Learn More
Memmert™ Iq/Oq Document W/Device-Specific Works Test Data (Temperature And Humidity Value), For Ich110Eco
Includes temperature distribution survey for one free-selectable temperature and humidity value.
Brand:  Memmert™ ICH110ecoD00136
Product Code. 16325227
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes:
Works test data, temperature distribution survey
Description
Description
  • IQ/OQ document with works test data for chamber
  • 27 measuring points to DIN 12880:2007-05, PQ check list as support for validation by customer (models HPP and ICH)
Specifications
Specifications
110, 260, 750 Models Constant Climate Chamber
IQ/OQ Documentation
One Free-Selectable Temperature and Humidity Value for 27 Measuring Points to DIN 12880:2007-05
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Iq/Oq Document W/Device-Specific Works Test Data (Temperature And Humidity Value), For Ich110Eco >