missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IQCH (Human) Recombinant Protein

Human IQCH full-length ORF ( NP_001026885.1, 1 a.a. - 1027 a.a.) recombinant protein with GST-tag at N-terminal.

3970.00 SEK - 6025.00 SEK

Specifications

Accession Number NP_001026885.1
Gene ID (Entrez) 64799
Name IQ motif containing H
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16133753
View Documents Promotion Details
Abnova™
H00064799-P01.10UG
10 μg
3970.00 SEK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 13-01-2023
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16143753
View Documents Promotion Details
Abnova™
H00064799-P01.25UG
25 μg
6025.00 SEK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 13-01-2023
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Sequence: MAQNTENHDPVGSILIQIHEDLYQLKEKLTKFSPEEKGETLDIQSLETAIKRTEVGLRIHIEKYLNVVNQNVLTTSVNDESLYTPQASKWLLPTVIDQKSFIFPQESEGTFWQPQRQHSSSLPVFPRAKIKVSKLIKGSNISSLTVLPSSHCTDPYFTPIPVLQADAHKGILSMIERGLIPPTARITFQNPPITPRAAPLHSFDEARKIPTVATFTIPREPPPSPAEVKFFPKKQRSKGKSRRSRGHHDRKAMKVKTPLRALKSLWDYDFLIYDGVIDNTAPDFLAFKEHFSLAWGGIFSLLEHVEKFLRNYAIPEVKIKGNNLVALLPEFELTNKLTRYDLLSVLEDPAHVQMLINLPGQRYKGQDGNSEAAMKIQATWKCYKARKFFLFYRQQKWASGVIAIAWLLYCHKTRLKKILKESRQRHLENFRIRAKHLAANWNRIRTSRRTIIHIPSLGYSQPVREHIADFNTQQNMQLGRLCDILDANVNVIYICSHHMNDELVLYYKKILSLHAAVKSGNLEDRSDLQDRFKIITPEAVNIFPKHHMCLATHLMYSPKAIKRIKNLIRGTEAYIVSGLLHRDDLAVADMLDIPILGSEPELAHLYSTKSGGKRVFDSANVAVPPGIYDIYSQQQMIEQLSQLITDHLQIQRWLFKMDSEFRGNGTAFCDIPSYLKCYKWVLKESSRYGLEDWRKKWAQEPALVKISEELAGILAQHAQPVNEKRFPTWRKFLQTFLSQGGVIEAFPPADNVTNLTVDMLIEPNGKISVLSTGDQLHAESPFISSGTTVPQTSVDPQVLTYLCLQIGKACRMRDVVGYFSIDLVTFIDPSTLEQQVWATGLNLAYSDQLALTQLTLYLTNGHLDCSLSTLEVPRFVPKERKKTKCMSALSMPMLATSRYAVMTTQLRHSNLSLVFHYVFLQICRAHGIGYDLEERQGTVFILYEHLKRHKLGMLTIGEDLQGVLMTFARHLFIIHQEISAPNMQGETNFKTTIADIETILRVTKENKMRFEEEQQSKDDKNLSKPKK
Specifications

Specifications

NP_001026885.1
IQ motif containing H
125% SDS-PAGE Stained with Coomassie Blue
DKFZp434F2114, FLJ12476, NYDSP5
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
64799
Wheat germ expression system
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IQCH
GST
SDS
Product Certifications