missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF2BP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13698-1-AP-20UL
This item is not returnable.
View return policy
Description
IRF2BP1 acts as a transcriptional repressor. IRF2BP1 also acts as a transcriptional corepressor in a IRF2-dependent manner. This repression is not mediated at least in part by histone deacetylase activities.Specifications
IRF2BP1 | |
Polyclonal | |
Unconjugated | |
IRF2BP1 | |
IRF 2 binding protein 1, IRF 2BP1, IRF2BP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26145, 272359, 308404 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunocytochemistry | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IU81, Q8R3Y8 | |
Irf2bp1 | |
IRF2BP1 Fusion Protein Ag4596 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IRF2BP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction