Learn More
Iron Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™ Product Code.: 10090190
Iron Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Product Code. 10090190 Shop All Fisher Chemical Products
500 mL, 500mL
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 10090190
Brand: Fisher Chemical J/8030/08
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Iron Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10421-48-4
Molecular Formula FeN3O9
Molecular Weight (g/mol) 241.857
MDL Number 11003
InChI Key VCJMYUPGQJHHFU-UHFFFAOYSA-N
Synonym ferric nitrate, iron trinitrate, nitric acid, iron 3+ salt, unii-n8h8402xob, iron nitrate fe no3 3, iron iii nitrate, anhydrous, hsdb 451, iron 3+ trinitrate, nitric acid, iron 3+ salt 3:1
PubChem CID 25251
IUPAC Name iron(3+);trinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Fe+3]
Specifications
Color Colorless
pH 1
Quantity 500 mL
Packaging HDPE plastic bottle
Form Liquid
Product Content Correction
Product Title
Iron Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™ >