missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPE Scientific™ J Young T Shape Taps, Fast Thread Type
GPE Scientific™ J Young T Shape Taps, Fast Thread Type are made in various stopcock sizes and with different side arm diameters.
Brand: GPE Scientific™ PTT/T/8
This item is not returnable.
View return policy
Description
- Fast thread type
- Available in various stopcock sizes
- Available with different side arm diameters
Specifications
J Young T Shape Tap, Fast Thread | |
8 mm | |
ID: 8 mm, OD: 11 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GPE Scientific™ J Young T Shape Taps, Fast Thread Type > Stopcock: 8mm
Spot an opportunity for improvement?Share a Content Correction