Learn More
Blue Eagle K-Series Safety Face Shields with Browguard and Visor
Click to view available options
Color:
Blue
Green
Orange
Yellow
Unit Size:
Pack of 100
Description
- Box containing 100 browguards with headband and sweatband as well as 100 visors
- Visor made of 0.8 mm thick polycarbonate plastic
- Requires assembly prior to first use
- Tested and approved for broader temperature working environment
- Available in range of different colors
- Certified as personal protective equipment
Compliance
This product complies with the essential requirements for health and safety as they are provided by the European Regulation (EU) 2016/425 for Personal Protective Equipment.
Specifications
Specifications
Lens Type | Highest optical class 1 |
Thickness (Metric) | 0.8 mm |
Description | Requires assembly prior to first use |
Color | Orange |
Certifications/Compliance (Europe) | Regulation (EU) 2016/425 |
Lens Tint | Clear |
For Use With (Application) | Approved for environmental temperature range -5 to +55°C |
Headband Material | Polypropylene/Polyester |
Quantity | 100/Cs |
Lens Material | Polycarbonate |
Show More |
Documents
Scientific Resources
Flyers
Safety Face Shield with Browguard and Visor
File size: 373.2 kB
Product Information
Instructions
Covid-19 Faceshield Assembly Instructions
File size: 155.7 kB
Product Information
EU Type Examination Certificate K4 K25N
File size: 132.3 kB
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Blue Eagle K-Series Safety Face Shields with Browguard and Visor
Spot an opportunity for improvement?Share a Content Correction