missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | Kallikrein 5 |
---|---|
Concentration | 0.43 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16896943
|
Proteintech
10514-2-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16886943
|
Proteintech
10514-2-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.Specifications
Kallikrein 5 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q9Y337 | |
25818 | |
Kallikrein 5 Fusion Protein Ag0799 | |
Primary | |
-20°C | |
KLK5 |
0.43 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Kallikrein 5, Kallikrein like protein 2, kallikrein related peptidase 5, KLK L2, KLK5, KLKL2, SCTE, Stratum corneum tryptic enzyme | |
KLK5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kallikrein 5 Rabbit anti-Human, Polyclonal, Proteintech