missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kanamycin monosulfate, MP Biomedicals™
876.00 SEK - 2535.00 SEK
Chemical Identifiers
CAS | 25389-94-0 |
---|---|
Molecular Formula | C18H38N4O15S |
Molecular Weight (g/mol) | 582.575 |
InChI Key | OOYGSFOGFJDDHP-IZQIRFRQSA-N |
Synonym | Kanamycin sulfate salt, Kanamycin A |
PubChem CID | 134129479 |
IUPAC Name | (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid |
SMILES | C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11466412
|
MP Biomedicals
0215002905 |
5g |
876.00 SEK
5g |
Estimated Shipment: 16-06-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11476412
|
MP Biomedicals
0215002925 |
25g |
2535.00 SEK
25g |
Estimated Shipment: 27-06-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
25389-94-0 | |
582.575 | |
Kanamycin sulfate salt, Kanamycin A | |
(2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid |
C18H38N4O15S | |
OOYGSFOGFJDDHP-IZQIRFRQSA-N | |
134129479 | |
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O |
Specifications
Kanamycin monosulfate | |
C18H38N4O15S | |
OOYGSFOGFJDDHP-IZQIRFRQSA-N | |
(2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid | |
134129479 | |
Powder | |
5g |
25389-94-0 | |
Kanamycin sulfate salt, Kanamycin A | |
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O | |
582.575 | |
582.6 | |
White |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Propidium Iodide
Signal Word
- Danger
Hazard Category
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H341-Suspected of causing genetic defects.
- H335-May cause respiratory irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.