Learn More
KCNH7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13622-1-AP-20UL
 View more versions of this product
Product Code. 16881924
  • 1645.54 SEK / 20µL
Description
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit. There are at least two alternatively spliced transcript variants derived from this gene and encoding distinct isoforms.
Specifications
Specifications
KCNH7
Polyclonal
Unconjugated
KCNH7
Eag related protein 3, ERG 3, ERG3, hERG 3, HERG3, KCNH7, Kv11.3
Rabbit
Antigen Affinity Chromatography
RUO
170738, 170739, 90134
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O54852, Q9ER47, Q9NS40
KCNH7
KCNH7 Fusion Protein Ag4532
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
KCNH7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated