missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNV1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12684-1-AP-150UL
This item is not returnable.
View return policy
Description
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium voltage-gated channel subfamily V. This protein is essentially present in the brain, and its role might be to inhibit the function of a particular class of outward rectifier potassium channel types.Specifications
KCNV1 | |
Polyclonal | |
Unconjugated | |
KCNV1 | |
HNKA, KCNB3, KCNV1, KV2.3, KV8.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27012, 60326, 67498 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P97557, Q6PIU1, Q8BZN2 | |
KCNV1 | |
KCNV1 Fusion Protein Ag3376 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
KCNV1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction