Learn More
KDM4D Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22591-1-AP-150UL
 View more versions of this product
Product Code. 16881995
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
JMJD2D belongs to JMJD2 family that is consist of four cancer-associated genes.
Specifications
Specifications
KDM4D
Polyclonal
Unconjugated
KDM4D
JHDM3D, JMJD2D, KDM4D, lysine (K) demethylase 4D, Lysine specific demethylase 4D
Rabbit
Antigen Affinity Chromatography
RUO
55693
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6B0I6
KDM4D
KDM4D Fusion Protein Ag18436
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
KDM4D Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated