Learn More
DWK Life Sciences KECK™ Suction Connector, AS M8, complete with tubing parts
For suction or supply of liquids and gases.
67.46 SEK - 1465.00 SEK
Specifications
Quantity | 10 |
---|---|
For Use With (Equipment) | Extraction or Supply of Liquids and Gases From (or in) Cylinders, Containers, Tanks |
For Use With (Application) | Extraction or supply of liquids and gases from (or in) cylinders, containers, tanks |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15844698
|
DWK Life Sciences
286343001 |
4.5 mm |
1465.00 SEK
Pack of 10 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
10253591
|
DWK Life Sciences
286343104 |
9 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The KECK™ suction nozzle with hose part is usable for suction or supply of liquids and gases from (or in) cylinders, containers, tanks, through walls with an opening of Ø 8-11 mm. Available in 2 sizes: 4.5 mm and 9 mm.
- Suction nozzle with hose part for suction or supply of liquids and gases
- For hoses with a diameter of Ø 8-11 mm
- Available in 2 sizes: 4.5 mm and 9 mm.
Specifications
10 | |
Extraction or Supply of Liquids and Gases From (or in) Cylinders, Containers, Tanks | |
Extraction or supply of liquids and gases from (or in) cylinders, containers, tanks |