Learn More
KIF2C Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28372-1-AP-20UL
 View more versions of this product
Product Code. 16886115
  • 1645.54 SEK / 20µL
Description
The KIF2C antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KIF2C. This antibody recognizes human, mouse , rat antigen. The KIF2C antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
KIF2C
Polyclonal
Unconjugated
20 μL
11004
IgG
Western Blot, Immunohistochemistry, ELISA
300 μg/mL
Rabbit
Primary
Human, Rat, Mouse
Certificates
Promotions
Promotions
Product Content Correction
Product Title
KIF2C Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 20 μL