missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF5A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21186-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the kinesin family of proteins. Members of this family are part of a multisubunit complex that functions as a microtubule motor in intracellular organelle transport. Mutations in this gene cause autosomal domit spastic paraplegia 10.Specifications
KIF5A | |
Polyclonal | |
Unconjugated | |
KIF5A | |
D12S1889, KIF5A, kinesin family member 5A, Kinesin heavy chain isoform 5A, MY050, Neuronal kinesin heavy chain, NKHC, NKHC1, SPG10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16572, 314906, 3798 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P33175, Q12840, Q6QLM7 | |
Kif5a | |
KIF5A Fusion Protein Ag15518 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
KIF5A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction