Learn More
DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached PTFE-Faced/White Rubber Lined Caps
Packaged in corrugated trays with partitions
Brand: DWK Life Sciences 60811B-2
This item is not returnable.
View return policy
Description
- Small case quantities
- Manufactured from clear 33 expansion, borosilicate glass
- Attached closed top black phenolic closures
Specifications
A | |
Clear | |
Borosilicate Glass | |
Screw | |
Phenolic | |
60 mm |
PTFE/Rubber | |
17 mm | |
8 mL | |
2.16 dr. | |
Screw Cap (Closed-top) |
Promotions
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
Product Content Correction
Product Title
DWK Life Sciences Kimble™ ClearScrew Thread Sample Vials with Attached PTFE-Faced/White Rubber Lined Caps > Capacity: 2 dr.
Spot an opportunity for improvement?Share a Content Correction