Learn More
DWK Life Sciences Kimble™ Kontes™ ULTRA-WARE™ Heavy-Wall Filter Flasks
Heavy-wall borosilicate glass provides mechanical strength for vacuum filtration
Brand: DWK Life Sciences 953760-1002
This item is not returnable.
View return policy
Description
Heavy-wall borosilicate glass provides mechanical strength for vacuum filtration
Kimble Chase KimCote™ plastic-coated glassware provides an added measure of safety in the event of breakage
- KimCote™ plastic safety coating
- ≠2 side arm for connection to 0..25 in. (6mm) I.D. flexible tubing
- 125mL flask: With No. 5 stopper joint
- 1, 2 and 4L flasks: With No. 8 stopper joint
Specifications
0.93 cm | |
1000 mL | |
Borosilicate Glass | |
8 |
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Kontes™ ULTRA-WARE™ Heavy-Wall Filter Flasks > Capacity: 1L
Spot an opportunity for improvement?Share a Content Correction