missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19028-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Conventional kinesin is a tetrameric molecule composed of two heavy chains and two light chains, and transports various cargos along microtubules toward their plus ends. The heavy chains provide the motor activity, while the light chains bind to various cargos. This gene encodes a member of the kinesin light chain family.Specifications
KLC1 | |
Polyclonal | |
Unconjugated | |
KLC1 | |
kinesin light chain 1, KLC, KLC 1, KLC1, KNS2, KNS2A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16593, 171041, 3831 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88447, P37285, Q07866 | |
Klc1 | |
KLC1 Fusion Protein Ag13570 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |