missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13156-1-AP-20UL
This item is not returnable.
View return policy
Description
Activator protein-2 alpha (AP-2 alpha) is a developmentally-regulated transcription factor and important regulator of gene expression during vertebrate development and carcinogenesis. The protein encoded by this gene is a member of the Kruppel-like zinc finger protein family and can repress expression of the AP-2 alpha gene by binding to a specific site in the AP-2 alpha gene promoter. Repression by the encoded protein requires binding with a corepressor, CtBP1. Two transcript variants encoding different isoforms have been found for this gene.Specifications
KLF12 | |
Polyclonal | |
Unconjugated | |
KLF12 | |
AP 2rep, AP2REP, HSPC122, KLF12, Krueppel like factor 12, Kruppel like factor 12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11278, 16597, 306110 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35738, Q9Y4X4 | |
Klf12 | |
KLF12 Fusion Protein Ag3788 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
KLF12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction