missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KTN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19841-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody reacts with normal gastrointestinal epithelium as well as gastrointestinal and breast tumors, resulting in a cytoplasmic staining pattern.
This antibody reacts with normal gastrointestinal epithelium as well as gastrointestinal and breast tumors, resulting in a cytoplasmic staining pattern.Specifications
KTN1 | |
Polyclonal | |
Unconjugated | |
KTN1 | |
CG 1 antigen, CG1, KIAA0004, Kinectin, kinectin 1 (kinesin receptor), Kinesin receptor, KNT, KTN1, MU RMS 40.19 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16709, 3895 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q61595, Q86UP2 | |
KTN1 | |
KTN1 Fusion Protein Ag13854 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
KTN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction