missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv1.3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14079-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
In neurons voltage-dependent potassium channels are key determits of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.Specifications
Kv1.3 | |
Polyclonal | |
Unconjugated | |
KCNA3 | |
HGK5, HLK3, HPCN3, HUKIII, KCNA3, Kv1.3, MK3, PCN3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16491, 3738 | |
-20°C | |
Liquid |
Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P16390, P22001 | |
Kcna3 | |
Kv1.3 Fusion Protein Ag5204 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |