missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv1.3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | Kv1.3 |
---|---|
Concentration | 0.21 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16832564
|
Proteintech
14079-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16822564
|
Proteintech
14079-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
In neurons voltage-dependent potassium channels are key determits of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.Specifications
Kv1.3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P16390, P22001 | |
16491, 3738 | |
Kv1.3 Fusion Protein Ag5204 | |
Primary | |
-20°C | |
KCNA3 |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HGK5, HLK3, HPCN3, HUKIII, KCNA3, Kv1.3, MK3, PCN3 | |
Kcna3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |