missing translation for 'onlineSavingsMsg'
Learn More

Kv1.3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14079-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16822564

  • 4721.67 SEK / 150µL
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

In neurons voltage-dependent potassium channels are key determits of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.
Specifications

Specifications

Kv1.3
Polyclonal
Unconjugated
KCNA3
HGK5, HLK3, HPCN3, HUKIII, KCNA3, Kv1.3, MK3, PCN3
Rabbit
Antigen Affinity Chromatography
RUO
16491, 3738
-20°C
Liquid
Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P16390, P22001
Kcna3
Kv1.3 Fusion Protein Ag5204
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers