Learn More
Masterflex™ L/S™ Standard Pump Head Replacement End Bell Assembly
Masterflex™ L/S™ Standard Pump Head Replacement End Bell Assemblies extend the operating life of standard pump heads
Brand: Masterflex™ 07018-81
1294.05 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Replacement parts to keep pump head operating for an extended period of time
- Two end bell assemblies are required for a complete head assembly
- Choose replacement parts suitable for your I/P™ Standard pump head size
- Suitable for use with L/S™ Standard Pump Heads
Specifications
CRS Rotor, L/S 18 Tubing | |
L/S™ Standard Pump Heads 07018-00, 07018-20, 07018-70, and 07018-72 | |
Replacement End Bell Assembly |
Polycarbonate | |
L/S 18 |
Product Content Correction
Product Title
Masterflex™ L/S™ Standard Pump Head Replacement End Bell Assembly > CRS Rotor, L/S 18 Tubing
Spot an opportunity for improvement?Share a Content Correction