Learn More
Hirschmann™ Labopette™ Variable Volume Single Channel Manual Pipette
Ergonomically shaped manual pipette, effortless operation and tip ejector.
Brand: Hirschmann™ 9475409
2850.95 SEK valid until 2025-07-31
Use promo code "24075" to get your promotional price.
This item is not returnable.
View return policy
Description
- Variable volume setting
- With tip ejector and recalibration possibility
- Autoclavable
Specifications
0.8% | |
10 to 1000 ÎĽL | |
Yellow | |
Labopette | |
100 to 1000 ÎĽL | |
1 ÎĽL |
CE, IVD 98/79 EC | |
Autoclavable | |
1 | |
Labopettor | |
Calibration tool and pipette holder, individual serial number and calibration certificate |
Product Content Correction
Product Title
Hirschmann™ Labopette™ Variable Volume Single Channel Manual Pipette > Volume: 100 to 1000 μL
Spot an opportunity for improvement?Share a Content Correction