Läs mer
Invitrogen™ LanthaScreen™ Tb-pSer (PKC) Antibody
A general assay for the Protein Kinase C (PKC) family, the antibody detects the phosphorylation of the serine within the PKC substrate.
Brand: Invitrogen™ PV3537
Beskrivning
A general assay for the Protein Kinase C (PKC) family, the antibody detects the phosphorylation of the serine within the PKC substrate.
For use in combination with the Fluorescein-PKC substrate (Cat. No. PV3506) to evaluate inhibitors in medium to high throughput screening applications.
Specifikationer
| Tb (Terbium)/Lanthanide Chelates | |
| Terbium-labeled anti-pSer (PKC) antibody in HEPES buffered saline. Store at -20°C. |
|
| Human | |
| End Point | |
| Ser/Thr Kinases |
| 1 mg | |
| 340/490, 520 nm | |
| Dry Ice | |
| Hu | |
| LanthaScreen |