Läs mer
Invitrogen™ LanthaScreen™ Tb-pSer (PKC) Antibody
A general assay for the Protein Kinase C (PKC) family, the antibody detects the phosphorylation of the serine within the PKC substrate.
Brand: Invitrogen™ PV3536
Beskrivning
A general assay for the Protein Kinase C (PKC) family, the antibody detects the phosphorylation of the serine within the PKC substrate.
For use in combination with the Fluorescein-PKC substrate (Cat. No. PV3506) to evaluate inhibitors in medium to high throughput screening applications.
Specifikationer
| Serine/threonine kinase | |
| 1mg/mL | |
| 150kDa | |
| RUO | |
| Primary | |
| Terbium-labeled anti-pSer (PKC) antibody in HEPES buffered saline. Store at -20°C. |
|
| Human | |
| End Point | |
| Ser/Thr Kinases |
| Kinase Assay | |
| Tb (Terbium)/Lanthanide Chelates | |
| 25 μg | |
| Target and Lead Identification and Validation | |
| 1 Each | |
| 340/490, 520 nm | |
| Dry Ice | |
| Hu | |
| LanthaScreen |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
For Research Use Only. Not for use in diagnostic procedures.