Läs mer
(+)-Lappaconitine >90%, TRC
Produktkod. 30465092
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30465092 250 mg
30480092 500 mg
30477303 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30465092 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL175850250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 32854-75-4
Molekylformel C32 H44 N2 O8
Molekylvikt (g/mol) 584.7
Synonym (+)-Lappaconitine, Acetyl-10-deoxysepaconitine
LEDER CCN1C[C@@]2(CC[C@H](OC)C34[C@@H]2CC(C13)[C@@]5(O)C[C@H](OC)[C@H]6C[C@@H]4[C@]5(O)[C@H]6OC)OC(=O)c7ccccc7NC(=O)C
Specifikationer
Kemiskt namn eller material Lappaconitine
Kvantitet 250 mg
InChI formel InChI=1S/C32H44N2O8/c1-6-34-16-29(42-28(36)18-9-7-8-10-21(18)33-17(2)35)12-11-25(40-4)31-23(29)14-20(26(31)34)30(37)15-22(39-3)19-13-24(31)32(30,38)27(19)41-5/h7-10,19-20,22-27,37-38H,6,11-16H2,1-5H3,(H,33,35)/t19-,20?,22+,23-,24+,25+,26?,27+,29-,30+,31?,32+/m1/s1
Formel vikt 584.3098