missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDLR Mouse anti-Human, Clone: 1A2A3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66414-1-IG-150UL
This item is not returnable.
View return policy
Description
The low density lipoprotein (LDL) receptor system coordinates the metabolism of cholesterol, an essential component of the plasma membrane of all mammalian cells. Study of this system has led to an enhanced understanding of the cellular basis of cholesterol homeostasis. It has also brought into focus an important mechanism of metabolic regulation-the process of receptor-mediated endocytosis. Data suggest that the juxtamembranous region of the cytoplasmic domain participates in protein:protein interactions that allow the low density lipoprotein receptor to cluster in coated pits. It has been shown that the family of LDL receptors may serve as viral receptors. Endocytosis of the Flaviviridae viruses, hepatitis C virus, GB virus C/hepatitis G virus, and bovine viral diarrheal virus (BVDV) was shown to be mediated by LDL receptors on cultured cells.Specifications
LDLR | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P01130 | |
LDLR | |
LDLR Fusion Protein Ag1236 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1A2A3 | |
Unconjugated | |
LDLR | |
FH, FHC, LDL receptor, LDLR | |
Mouse | |
Protein G | |
RUO | |
3949 | |
-20°C | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LDLR Mouse anti-Human, Clone: 1A2A3, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction