Läs mer
Brand™ LDPE Wash Bottles without Valve
Handla allt Brand Produkter
Kapacitet (metrisk):
250 mL
500 mL
Kemisk etikett:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Neutral
Stängningsfärg:
Blue
Green
Orange
Red
White
Yellow
Förpackningsstorlek:
Paket med 5
Beskrivning
- Manufactured from flexible LDPE
- Wide-mouth.
- High-quality wash bottles with integrated wash stream and sealing cone closure.
- Large opening for convenient filling.
- Label with designation of contents and general safety statements in English (with CAS numbers and NFPA codes) and German (with CAS numbers).
Specifikationer
Specifikationer
| Kapacitet (metrisk) | 250 mL |
| Material | LDPE |
| Stängningsfärg | White |
| Mun | Wide |
| Produkttyp | Wash Bottle |
| Kemisk etikett | Distilled Water |
| Förslutningstyp | Sealing Cone |