Learn More
Bel-Art™ SP Scienceware™ Lead Sheets with Vikem Vinyl Coating
Soft lead sheets for stabilizing laboratory equipment in water baths or any application where extra weight is needed
Brand:  Bel-Art™ SP Scienceware™ F18321-0027
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11731963
  • 299.00 SEK / Each
Estimated Shipment
14-06-2021
Add to basket
Description
Description
  • Sheets may be bent into cylinders, S-curves, U- curves or twists to hold down various container types
  • Vikem™ vinyl coating prevents the lead from leaving marks on equipment
  • Coated lead sheets are 2.2mm (0.085 in.) thick
Specifications
Specifications
Stabilizing laboratory equipment
2 in.
Vinyl Coated Lead
Lead Sheet
Water bath
5.1 cm
2 x 7 in.