Learn More
LEPROT Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20304-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The LEPROT antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human LEPROT. This antibody recognizes human, pig antigen. The LEPROT antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
LEPROT | |
Polyclonal | |
Unconjugated | |
150 μL | |
54741 | |
IgG |
Western Blot, ELISA | |
367 μg/mL | |
Rabbit | |
Primary | |
Human, Pig |