missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LEPROT Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | LEPROT |
---|---|
Concentration | 367 μg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879824
|
Proteintech
20304-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16869824
|
Proteintech
20304-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The LEPROT antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human LEPROT. This antibody recognizes human, pig antigen. The LEPROT antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
LEPROT | |
Western Blot, ELISA | |
Unconjugated | |
Human, Pig | |
IgG |
367 μg/mL | |
Polyclonal | |
Rabbit | |
54741 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LEPROT Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech